Assay Details
Gene Symbol: Sra-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142190.3 | NP_650447.1 | 5 - 6 | 2763 | 103 | |
GenBank mRNA | AY017343.1 | - | 5 - 6 | 2750 | 103 | |
AY029211.1 | - | 5 - 6 | 2750 | 103 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
