Assay Details
Gene Symbol: Sra-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142190.3 | NP_650447.1 | 7 - 8 | 3397 | 73 | |
GenBank mRNA | AY017343.1 | - | 7 - 8 | 3384 | 73 | |
AY029211.1 | - | 7 - 8 | 3384 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
