Assay Details
Gene Symbol: Rbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170149.3 | NP_001163620.3 | 2 - 3 | 503 | 61 | |
NM_001202297.3 | NP_001189226.1 | 2 - 3 | 503 | 61 | ||
NM_001202298.3 | NP_001189227.1 | 2 - 3 | 503 | 61 | ||
NM_001275672.2 | NP_001262601.1 | 2 - 3 | 1212 | 61 | ||
NM_001275673.2 | NP_001262602.1 | 2 - 3 | 857 | 61 | ||
NM_001316579.1 | NP_001303508.1 | 2 - 3 | 503 | 61 | ||
NM_169652.5 | NP_732028.4 | 2 - 3 | 503 | 61 | ||
NM_169653.4 | NP_732029.4 | 2 - 3 | 503 | 61 | ||
NM_169654.4 | NP_732030.4 | 2 - 3 | 1212 | 61 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
