Assay Details
Gene Symbol: Atx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300410.1 | NP_001287339.1 | 3 - 4 | 1350 | 79 | |
NM_142209.3 | NP_650466.2 | 3 - 4 | 1414 | 79 | ||
NM_169657.3 | NP_732033.1 | 3 - 4 | 2097 | 79 | ||
NM_169658.3 | NP_732034.2 | 3 - 4 | 1414 | 79 | ||
GenBank mRNA | AY095516.1 | - | 3 - 4 | 1849 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
