Assay Details
| Gene Symbol: Acyp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275683.1 | NP_001262612.1 | 2 - 3 | 236 | 68 | |
| NM_142234.2 | NP_650491.1 | 2 - 3 | 236 | 68 | ||
| GenBank mRNA | BT024404.1 | - | 2 - 3 | 231 | 68 | |
More Information
Set Membership: |
|

Acyp2
Genomic Map