Assay Details
| Gene Symbol: Arpc3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142241.2 | NP_650498.3 | 2 - 3 | 300 | 90 | |
| NM_169675.3 | NP_732075.3 | 2 - 3 | 300 | 90 | ||
| GenBank mRNA | BT132882.1 | - | 2 - 3 | 272 | 90 | |
More Information
Set Membership: |
|

Arpc3A
Genomic Map