Assay Details
| Gene Symbol: Gyc89Da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043253.1 | NP_001036718.1 | 2 | 1769 | 90 | |
| NM_001043254.2 | NP_001036719.1 | 2 | 1704 | 90 | ||
| GenBank mRNA | BT022818.1 | - | 2 | 1592 | 90 | |
| BT022843.1 | - | 2 | 1769 | 90 | ||
More Information
Set Membership: |
|

Gyc89Da
Genomic Map