Assay Details
| Gene Symbol: Cad89D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142297.4 | NP_650554.3 | 7 - 8 | 5552 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cad89D| Gene Symbol: Cad89D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142297.4 | NP_650554.3 | 7 - 8 | 5552 | 73 | |
Set Membership: |
|