Assay Details
Gene Symbol: CCT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202307.2 | NP_001189236.1 | 2 - 3 | 247 | 57 | |
NM_001202308.2 | NP_001189237.1 | 2 - 3 | 232 | 57 | ||
NM_142315.4 | NP_650572.2 | 2 - 3 | 188 | 57 | ||
NM_169727.2 | NP_732167.1 | 1 - 2 | 326 | 57 | ||
GenBank mRNA | AY089543.1 | - | 2 - 3 | 156 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
