Assay Details
| Gene Symbol: Abd-B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275719.2 | NP_001262648.1 | 4 - 5 | 932 | 113 | |
| NM_001316542.1 | NP_001303471.1 | 4 - 5 | 5856 | 113 | ||
| NM_001316543.1 | NP_001303472.1 | 2 - 3 | 2087 | 113 | ||
| NM_001316544.1 | NP_001303473.1 | 4 - 5 | 932 | 113 | ||
| NM_001316545.1 | NP_001303474.1 | 4 - 5 | 932 | 113 | ||
| NM_080157.3 | NP_524896.2 | 2 - 3 | 2087 | 113 | ||
| NM_142320.2 | NP_650577.1 | 4 - 5 | 750 | 113 | ||
| NM_169735.2 | NP_732180.1 | 4 - 5 | 923 | 113 | ||
| NM_169736.2 | NP_732181.1 | 4 - 5 | 932 | 113 | ||
| NM_206498.2 | NP_996220.1 | 4 - 5 | 918 | 113 | ||
| GenBank mRNA | BT124750.1 | - | 4 - 5 | 928 | 113 | |
More Information
Set Membership: |
|

Abd-B
Genomic Map