Assay Details
| Gene Symbol: AdSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142329.4 | NP_650586.2 | 1 | 2200 | 99 | |
| GenBank mRNA | BT044196.1 | - | 1 | 2155 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AdSL| Gene Symbol: AdSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142329.4 | NP_650586.2 | 1 | 2200 | 99 | |
| GenBank mRNA | BT044196.1 | - | 1 | 2155 | 99 | |
Set Membership: |
|