Assay Details
Gene Symbol: Patr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300440.1 | NP_001287369.1 | 4 - 5 | 2716 | 108 | |
NM_142335.3 | NP_650592.1 | 4 - 5 | 2716 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
