Assay Details
Gene Symbol: Ns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169742.3 | NP_732199.2 | 2 - 3 | 974 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
