Assay Details
Gene Symbol: Keap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142337.3 | NP_650594.1 | 4 - 5 | 1774 | 65 | |
NM_169744.3 | NP_732202.2 | 4 - 5 | 1536 | 65 | ||
NM_176508.3 | NP_788685.1 | 4 - 5 | 1324 | 65 | ||
GenBank mRNA | BT099945.1 | - | 4 - 5 | 1532 | 65 | |
DQ372684.1 | - | 4 - 5 | 1400 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
