Assay Details
Gene Symbol: Pxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142391.5 | NP_650648.3 | 4 - 5 | 1824 | 59 | |
GenBank mRNA | AY119616.1 | - | 4 - 5 | 1764 | 59 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
