Assay Details
| Gene Symbol: Rim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260231.1 | NP_001247160.1 | 17 - 18 | 6458 | 93 | |
| NM_001260234.1 | NP_001247163.1 | 17 - 18 | 6458 | 93 | ||
| NM_001260239.1 | NP_001247168.1 | 17 - 18 | 6458 | 93 | ||
More Information
Set Membership: |
|

Rim
Genomic Map