Assay Details
| Gene Symbol: Rim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014630.4 | NP_001014630.3 | 5 - 6 | 2007 | 75 | |
| NM_001260231.1 | NP_001247160.1 | 5 - 6 | 2007 | 75 | ||
| NM_001260232.2 | NP_001247161.2 | 5 - 6 | 2007 | 75 | ||
| NM_001260233.2 | NP_001247162.2 | 5 - 6 | 2007 | 75 | ||
| NM_001260234.1 | NP_001247163.1 | 5 - 6 | 2007 | 75 | ||
| NM_001260235.2 | NP_001247164.2 | 5 - 6 | 2007 | 75 | ||
| NM_001260237.2 | NP_001247166.2 | 5 - 6 | 2007 | 75 | ||
| NM_001260238.2 | NP_001247167.2 | 5 - 6 | 2007 | 75 | ||
| NM_001260239.1 | NP_001247168.1 | 5 - 6 | 2007 | 75 | ||
| NM_001260240.2 | NP_001247169.2 | 5 - 6 | 2007 | 75 | ||
| NM_001260241.2 | NP_001247170.2 | 5 - 6 | 2007 | 75 | ||
| NM_001260242.2 | NP_001247171.2 | 5 - 6 | 2007 | 75 | ||
| NM_001260243.2 | NP_001247172.2 | 5 - 6 | 2007 | 75 | ||
| NM_001275758.1 | NP_001262687.1 | 5 - 6 | 2007 | 75 | ||
More Information
Set Membership: |
|

Rim
Genomic Map