Assay Details
Gene Symbol: GluClalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014641.4 | NP_001014641.2 | 2 - 3 | 717 | 132 | |
NM_001038972.4 | NP_001034061.2 | 2 - 3 | 717 | 132 | ||
NM_001170183.2 | NP_001163654.1 | 2 - 3 | 717 | 132 | ||
NM_001170184.2 | NP_001163655.1 | 2 - 3 | 717 | 132 | ||
NM_001170185.3 | NP_001163656.2 | 2 - 3 | 717 | 132 | ||
NM_001275811.2 | NP_001262740.1 | 2 - 3 | 717 | 132 | ||
NM_001300479.1 | NP_001287408.1 | 2 - 3 | 717 | 132 | ||
NM_001300480.1 | NP_001287409.1 | 2 - 3 | 717 | 132 | ||
NM_001300481.1 | NP_001287410.1 | 2 - 3 | 717 | 132 | ||
NM_142570.4 | NP_650827.3 | 2 - 3 | 717 | 132 | ||
NM_169873.4 | NP_732447.2 | 2 - 3 | 717 | 132 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001170183.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
