Assay Details
Gene Symbol: GluClalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275811.2 | NP_001262740.1 | 9 - 10 | 1671 | 74 | |
NM_001300479.1 | NP_001287408.1 | 9 - 10 | 1668 | 74 | ||
NM_142570.4 | NP_650827.3 | 9 - 10 | 1668 | 74 | ||
NM_169873.4 | NP_732447.2 | 9 - 10 | 1668 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
