Assay Details
| Gene Symbol: Ire1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104369.2 | NP_001097839.1 | 6 - 7 | 2628 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ire1| Gene Symbol: Ire1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104369.2 | NP_001097839.1 | 6 - 7 | 2628 | 107 | |
Set Membership: |
|