Assay Details
| Gene Symbol: CG4538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300484.1 | NP_001287413.1 | 2 - 3 | 2233 | 75 | |
| NM_142586.2 | NP_650843.1 | 2 - 3 | 1779 | 75 | ||
| NM_169880.2 | NP_732463.2 | 2 - 3 | 2233 | 75 | ||
| GenBank mRNA | BT032831.1 | - | 2 - 3 | 1779 | 75 | |
More Information
Set Membership: |
|

CG4538
Genomic Map