Assay Details
| Gene Symbol: Lrrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104377.2 | NP_001097847.1 | 6 - 7 | 4338 | 59 | |
| NM_001275842.1 | NP_001262771.1 | 6 - 7 | 4338 | 59 | ||
| NM_001275843.1 | NP_001262772.1 | 7 - 8 | 4611 | 59 | ||
| GenBank mRNA | DQ494533.1 | - | 6 - 7 | 4249 | 59 | |
More Information
Set Membership: |
|

Lrrk
Genomic Map