Assay Details
| Gene Symbol: Rlip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275863.1 | NP_001262792.1 | 1 - 2 | 192 | 67 | |
| NM_142676.4 | NP_650933.2 | 1 - 2 | 192 | 67 | ||
More Information
Set Membership: |
|

Rlip
Genomic Map