Assay Details
| Gene Symbol: Qsox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142741.4 | NP_650998.1 | 1 | 1610 | 103 | |
| GenBank mRNA | AY070518.1 | - | 1 | 1584 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qsox2| Gene Symbol: Qsox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142741.4 | NP_650998.1 | 1 | 1610 | 103 | |
| GenBank mRNA | AY070518.1 | - | 1 | 1584 | 103 | |
Set Membership: |
|