Assay Details
| Gene Symbol: Sar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170000.3 | NP_732717.1 | 3 - 4 | 327 | 108 | |
| NM_170001.3 | NP_732718.1 | 3 - 4 | 625 | 108 | ||
| NM_170002.3 | NP_732719.1 | 3 - 4 | 376 | 108 | ||
| NM_206542.2 | NP_996265.1 | 2 - 3 | 276 | 108 | ||
| GenBank mRNA | BT001745.1 | - | 3 - 4 | 320 | 108 | |
More Information
Set Membership: |
|

Sar1
Genomic Map