Assay Details
| Gene Symbol: Sar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170000.3 | NP_732717.1 | 5 - 6 | 628 | 53 | |
| NM_170001.3 | NP_732718.1 | 5 - 6 | 926 | 53 | ||
| NM_170002.3 | NP_732719.1 | 5 - 6 | 677 | 53 | ||
| NM_206542.2 | NP_996265.1 | 4 - 5 | 577 | 53 | ||
| GenBank mRNA | BT001745.1 | - | 5 - 6 | 621 | 53 | |
More Information
Set Membership: |
|

Sar1
Genomic Map