Assay Details
| Gene Symbol: AP-2mu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170215.2 | NP_001163686.1 | 2 - 3 | 760 | 65 | |
| NM_142792.5 | NP_651049.3 | 2 - 3 | 760 | 65 | ||
| NM_170015.3 | NP_732744.1 | 3 - 4 | 700 | 65 | ||
| GenBank mRNA | AF110232.1 | - | 2 - 3 | 655 | 65 | |
More Information
Set Membership: |
|

AP-2mu
Genomic Map