Assay Details
| Gene Symbol: Nup133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142823.3 | NP_651080.2 | 6 - 7 | 2780 | 60 | |
| GenBank mRNA | BT003226.1 | - | 6 - 7 | 2859 | 60 | |
More Information
Set Membership: |
|

Nup133
Genomic Map