Assay Details
| Gene Symbol: Ublcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142861.4 | NP_651118.1 | 1 - 2 | 320 | 73 | |
| GenBank mRNA | AF132171.1 | - | 1 - 2 | 300 | 73 | |
More Information
Set Membership: |
|

Ublcp1
Genomic Map