Assay Details
| Gene Symbol: bb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142883.2 | NP_651140.1 | 1 - 2 | 474 | 77 | |
| GenBank mRNA | BT015308.1 | - | 1 - 2 | 469 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bb8| Gene Symbol: bb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142883.2 | NP_651140.1 | 1 - 2 | 474 | 77 | |
| GenBank mRNA | BT015308.1 | - | 1 - 2 | 469 | 77 | |
Set Membership: |
|