Assay Details
Gene Symbol: Irk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300556.1 | NP_001287485.1 | 2 - 3 | 546 | 97 | |
NM_142892.2 | NP_651149.2 | 2 - 3 | 546 | 97 | ||
GenBank mRNA | AY051445.1 | - | 2 - 3 | 546 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
