Assay Details
| Gene Symbol: Irk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300556.1 | NP_001287485.1 | 5 - 6 | 1711 | 74 | |
| NM_142892.2 | NP_651149.2 | 5 - 6 | 1708 | 74 | ||
| NM_170076.2 | NP_732871.1 | 4 - 5 | 1414 | 74 | ||
| GenBank mRNA | AY051445.1 | - | 5 - 6 | 1708 | 74 | |
More Information
Set Membership: |
|

Irk2
Genomic Map