Assay Details
Gene Symbol: SPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142911.4 | NP_651168.1 | 1 - 2 | 106 | 60 | |
GenBank mRNA | AY051638.1 | - | 1 - 2 | 91 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
