Assay Details
| Gene Symbol: Rpn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142920.3 | NP_651177.1 | 3 - 4 | 851 | 96 | |
| NM_170088.2 | NP_732899.1 | 4 - 5 | 961 | 96 | ||
| GenBank mRNA | AF145311.1 | - | 3 - 4 | 745 | 96 | |
| AY061232.1 | - | 3 - 4 | 836 | 96 | ||
More Information
Set Membership: |
|

Rpn9
Genomic Map