Assay Details
Gene Symbol: SdhD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142924.3 | NP_651181.1 | 3 - 4 | 460 | 68 | |
GenBank mRNA | AY071388.1 | - | 3 - 4 | 457 | 68 | |
AY071617.1 | - | 3 - 4 | 457 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
