Assay Details
Gene Symbol: CG13604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104428.1 | NP_001097898.1 | 1 - 2 | 238 | 81 | |
NM_001275965.1 | NP_001262894.1 | 1 - 2 | 238 | 81 | ||
NM_142945.3 | NP_651202.1 | 1 - 2 | 238 | 81 | ||
GenBank mRNA | AY051411.1 | - | 1 - 2 | 238 | 81 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
