Assay Details
| Gene Symbol: CG13604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104426.1 | NP_001097896.1 | 1 - 2 | 264 | 68 | |
| NM_001275965.1 | NP_001262894.1 | 4 - 5 | 773 | 68 | ||
| NM_142945.3 | NP_651202.1 | 4 - 5 | 773 | 68 | ||
| GenBank mRNA | AY051411.1 | - | 4 - 5 | 773 | 68 | |
| BT003239.1 | - | 1 - 2 | 264 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT003239.1, NM_001104426.1 transcripts.
Set Membership: |
|

CG13604
Genomic Map