Assay Details
Gene Symbol: CG13604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104426.1 | NP_001097896.1 | 3 - 4 | 748 | 79 | |
NM_001104427.1 | NP_001097897.1 | 2 - 3 | 541 | 79 | ||
NM_001104428.1 | NP_001097898.1 | 5 - 6 | 1155 | 79 | ||
NM_001275965.1 | NP_001262894.1 | 6 - 7 | 1257 | 79 | ||
NM_142945.3 | NP_651202.1 | 6 - 7 | 1257 | 79 | ||
GenBank mRNA | AY051411.1 | - | 6 - 7 | 1257 | 79 | |
BT003239.1 | - | 3 - 4 | 748 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT003239.1, NM_001104426.1, NM_001104427.1 transcripts.
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
