Assay Details
Gene Symbol: CG13604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104426.1 | NP_001097896.1 | 4 - 5 | 1633 | 63 | |
NM_001104427.1 | NP_001097897.1 | 3 - 4 | 1426 | 63 | ||
NM_001104428.1 | NP_001097898.1 | 6 - 7 | 2040 | 63 | ||
NM_001275965.1 | NP_001262894.1 | 7 - 8 | 2142 | 63 | ||
NM_142945.3 | NP_651202.1 | 7 - 8 | 2142 | 63 | ||
GenBank mRNA | AY051411.1 | - | 7 - 8 | 2142 | 63 | |
BT003239.1 | - | 4 - 5 | 1633 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
