Assay Details
| Gene Symbol: Mettl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142947.4 | NP_651204.1 | 2 - 3 | 471 | 76 | |
| GenBank mRNA | AY113286.1 | - | 2 - 3 | 436 | 76 | |
More Information
Set Membership: |
|

Mettl3
Genomic Map