Assay Details
Gene Symbol: CG11089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202357.1 | NP_001189286.1 | 3 - 4 | 1586 | 86 | |
NM_143048.2 | NP_651305.1 | 4 - 5 | 1663 | 86 | ||
GenBank mRNA | BT011462.1 | - | 4 - 5 | 1640 | 86 |
Gene Symbol: CG31381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT030439.1 | - | 3 - 4 | 3411 | 86 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
