Assay Details
| Gene Symbol: Nmnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276013.2 | NP_001262942.1 | 3 - 4 | 688 | 92 | |
| NM_001369970.1 | NP_001356924.1 | 3 - 4 | 688 | 92 | ||
| NM_143058.1 | NP_651315.2 | 3 - 4 | 724 | 92 | ||
| NM_170185.1 | NP_733064.2 | 3 - 4 | 724 | 92 | ||
| GenBank mRNA | AY089411.1 | - | 3 - 4 | 651 | 92 | |
| BT030750.1 | - | 3 - 4 | 723 | 92 | ||
More Information
Set Membership: |
|

Nmnat
Genomic Map