Assay Details
| Gene Symbol: XNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300598.1 | NP_001287527.1 | 2 - 3 | 2135 | 77 | |
| NM_001300599.1 | NP_001287528.1 | 1 - 2 | 2199 | 77 | ||
| NM_143141.4 | NP_651398.1 | 1 - 2 | 2199 | 77 | ||
| NM_170228.3 | NP_733107.1 | 2 - 3 | 2135 | 77 | ||
| GenBank mRNA | AY058592.1 | - | 1 - 2 | 2437 | 77 | |
More Information
Set Membership: |
|

XNP
Genomic Map