Assay Details
Gene Symbol: XNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300598.1 | NP_001287527.1 | 3 - 4 | 3018 | 62 | |
NM_001300599.1 | NP_001287528.1 | 2 - 3 | 3082 | 62 | ||
NM_143141.4 | NP_651398.1 | 2 - 3 | 3082 | 62 | ||
NM_170228.3 | NP_733107.1 | 3 - 4 | 3018 | 62 | ||
GenBank mRNA | AY058592.1 | - | 2 - 3 | 3320 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
