Assay Details
| Gene Symbol: Npl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143150.4 | NP_651407.3 | 1 - 2 | 271 | 59 | |
| GenBank mRNA | AY058286.2 | - | 1 - 2 | 271 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npl4| Gene Symbol: Npl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143150.4 | NP_651407.3 | 1 - 2 | 271 | 59 | |
| GenBank mRNA | AY058286.2 | - | 1 - 2 | 271 | 59 | |
Set Membership: |
|