Assay Details
Gene Symbol: jigr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104450.2 | NP_001097920.1 | 5 - 6 | 948 | 86 | |
NM_001104451.2 | NP_001097921.1 | 3 - 4 | 948 | 86 | ||
NM_001276033.1 | NP_001262962.1 | 3 - 4 | 920 | 86 | ||
NM_001347754.1 | NP_001334702.1 | 5 - 6 | 1149 | 86 | ||
NM_143152.4 | NP_651409.1 | 5 - 6 | 1606 | 86 | ||
NM_170233.2 | NP_733112.1 | 5 - 6 | 1149 | 86 | ||
NM_170234.1 | NP_733113.1 | 5 - 6 | 1080 | 86 | ||
NM_170235.3 | NP_733114.1 | 4 - 5 | 886 | 86 | ||
GenBank mRNA | AY075409.1 | - | 5 - 6 | 1542 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
