Assay Details
Gene Symbol: ALiX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143325.4 | NP_651582.1 | 5 - 6 | 2525 | 61 | |
GenBank mRNA | AY069534.1 | - | 5 - 6 | 2623 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
