Assay Details
| Gene Symbol: CG12877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038984.3 | NP_001034073.1 | 2 - 3 | 1647 | 95 | |
| GenBank mRNA | BT133134.1 | - | 2 - 3 | 1645 | 95 | |
More Information
Set Membership: |
|

CG12877
Genomic Map