Assay Details
| Gene Symbol: mino | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260414.2 | NP_001247343.2 | 1 - 2 | 332 | 75 | |
| NM_143340.4 | NP_651597.1 | 1 - 2 | 806 | 75 | ||
| GenBank mRNA | AF145635.1 | - | 1 - 2 | 794 | 75 | |
| BT124983.1 | - | 1 - 2 | 532 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT124983.1 transcripts.
Set Membership: |
|

mino
Genomic Map