Assay Details
| Gene Symbol: mino | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260414.2 | NP_001247343.2 | 7 - 8 | 2558 | 68 | |
| NM_143340.4 | NP_651597.1 | 7 - 8 | 3032 | 68 | ||
| NM_170358.2 | NP_733237.1 | 6 - 7 | 2389 | 68 | ||
| NM_170359.3 | NP_733238.1 | 6 - 7 | 2278 | 68 | ||
| GenBank mRNA | AF145635.1 | - | 7 - 8 | 3020 | 68 | |
| BT124983.1 | - | 8 - 9 | 3248 | 68 | ||
More Information
Set Membership: |
|

mino
Genomic Map